CAS RN 1454835-54-1
CAS RN 1454835-54-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 6,12-Dihydro-6,6,12,12-tetrakis(4-octylphenyl)-2,8-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)dithieno[2,3-d:2',3'-d']-s-indaceno[1,2-b:5,6-b']dithiophene
Sigma Aldrich 6,12-Dihydro-6,6,12,12-tetrakis(4-octylphenyl)-2,8-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)dithieno[2,3-d:2',3'-d']-s-indaceno[1,2-b:5,6-b']dithiophene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.