accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1455-20-5

IUPAC Name: 2-butylthiophene
Synonyms: thiophene, 2-butyl-
Molecular Weight (g/mol): 140.24
Molecular Formula: C8H12S
InChi Key: MNDZHERKKXUTOE-UHFFFAOYSA-N
SMILES: CCCCC1=CC=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results