accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHEr

CAS RN 14553-08-3

IUPAC Name: erbium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: erbium(3+) triacetylacetonate
Molecular Weight (g/mol): 464.59
Molecular Formula: C15H21ErO6
InChi Key: FGQSJRDKBCVFHH-UHFFFAOYSA-N
SMILES: [Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results