accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFONCO

CAS RN 14565-32-3

IUPAC Name: trifluoroethanecarbonyl isocyanate
Synonyms: trifluoroethanecarbonyl isocyanate
Molecular Weight (g/mol): 139.03
Molecular Formula: C3F3NO2
InChi Key: GDCXCDOQEWMQSO-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results