accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHN

CAS RN 1457-85-8

IUPAC Name: ethyl (phenylcarbamoyl)formate
Synonyms: oxanilic acid,ethyl ester
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: YDGAUBHNAKCSKF-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results