accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 145764-54-1

IUPAC Name: 5-[7,12,17-tris(3,5-dihydroxyphenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaen-2-yl]benzene-1,3-diol
Synonyms: 5-[7,12,17-tris(3,5-dihydroxyphenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaen-2-yl]benzene-1,3-diol
Molecular Weight (g/mol): 742.74
Molecular Formula: C44H30N4O8
InChi Key: XDVQXKKXBXLGCO-UHFFFAOYSA-N
SMILES: OC1=CC(=CC(O)=C1)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=CC(O)=CC(O)=C1)C1=CC(O)=CC(O)=C1)C1=CC(O)=CC(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results