accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3ClHNSiCH3H3CCl

CAS RN 14579-91-0

IUPAC Name: bis[(chloromethyl)dimethylsilyl]amine
Synonyms: bis[(chloromethyl)dimethylsilyl]amine
Molecular Weight (g/mol): 230.28
Molecular Formula: C6H17Cl2NSi2
InChi Key: SXSNZRHGAMVNJE-UHFFFAOYSA-N
SMILES: C[Si](C)(CCl)N[Si](C)(C)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results