accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 145819-99-4

IUPAC Name: 2-({2-[bis(carboxymethyl)amino]cyclohexyl}(carboxymethyl)amino)acetic acid hydrate
Synonyms: CDTA hydrate
Molecular Weight (g/mol): 364.35
Molecular Formula: C14H24N2O9
InChi Key: VASZYFIKPKYGNC-UHFFFAOYNA-N
SMILES: O.OC(=O)CN(CC(O)=O)C1CCCCC1N(CC(O)=O)CC(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results