accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHNd

CAS RN 14589-38-9

IUPAC Name: neodymium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: neodymium(3+) triacetylacetonate
Molecular Weight (g/mol): 441.57
Molecular Formula: C15H21NdO6
InChi Key: AZTDRGMPTCCVFW-UHFFFAOYSA-N
SMILES: [Nd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results