accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 1459-09-2

IUPAC Name: hexadecylbenzene
Synonyms: N-hexadecylbenzene
Molecular Weight (g/mol): 302.55
Molecular Formula: C22H38
InChi Key: DEQLTFPCJRGSHW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results