accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH3C

CAS RN 1459-93-4

IUPAC Name: 1,3-dimethyl benzene-1,3-dicarboxylate
Synonyms: dimethyl isophthalate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: VNGOYPQMJFJDLV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC=C1)C(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (9)
Percent Purity 
  • (2)
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (1)
  • (2)
  • (6)
  • (3)
  • (9)
  • (2)
  • (3)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)