accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14590-13-7

IUPAC Name: λ²-cobalt(2+) ammonium phosphate
Synonyms: λ²-cobalt(2+) ammonium phosphate
Molecular Weight (g/mol): 171.94
Molecular Formula: CoH4NO4P
InChi Key: FCXHHGTWMFGYPP-UHFFFAOYSA-L
SMILES: [NH4+].[Co++].[O-]P([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results