accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 146-14-5

IUPAC Name: {[5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}({[(5-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}-2,3,4-trihydroxypentyl)oxy](hydroxy)phosphoryl}oxy)phosphinic acid
Synonyms: flavin adenine dinucleotide disodium flavine adenine dinucleotide
Molecular Weight (g/mol): 785.56
Molecular Formula: C27H33N9O15P2
InChi Key: VWWQXMAJTJZDQX-UHFFFAOYNA-N
SMILES: CC1=C(C)C=C2N(CC(O)C(O)C(O)COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results