accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOH

CAS RN 1460-38-4

IUPAC Name: 2-(2-oxocyclopentyl)acetic acid
Synonyms: (2-oxocyclopentyl)acetic acid
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: OLLLIBGOZUPLOK-UHFFFAOYNA-N
SMILES: OC(=O)CC1CCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results