accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOH3CCH3H3CH3CCH3OHO

CAS RN 146000-39-7

IUPAC Name: 2-{[(tert-butoxy)carbonyl](methyl)amino}-2-methylpropanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)(methyl)amino]-2-methylpropanoic acid|BOC-N-ME-AIB-OH
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: KPDLITGXUYMJEC-UHFFFAOYSA-N
SMILES: CN(C(=O)OC(C)(C)C)C(C)(C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)