accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CONOOCH3

CAS RN 146039-03-4

IUPAC Name: 1,1-diethyl-3-formyl-3-methoxyurea
Synonyms: 1,1-diethyl-3-formyl-3-methoxyurea
Molecular Weight (g/mol): 174.2
Molecular Formula: C7H14N2O3
InChi Key: ABUWPGZRKFDPIX-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)N(OC)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results