accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3(R)NCH3CHH

CAS RN 14611-52-0

IUPAC Name: hydrogen methyl[(2R)-1-phenylpropan-2-yl](prop-2-yn-1-yl)amine chloride
Synonyms: selegiline
Molecular Weight (g/mol): 223.74
Molecular Formula: C13H18ClN
InChi Key: IYETZZCWLLUHIJ-UTONKHPSSA-N
SMILES: [H+].[Cl-].C[C@H](CC1=CC=CC=C1)N(C)CC#C
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)