accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 14615-72-6

IUPAC Name: 3,5-bis(benzyloxy)benzaldehyde
Synonyms: 3,5-bis(benzyloxy)benzaldehyde
Molecular Weight (g/mol): 318.37
Molecular Formula: C21H18O3
InChi Key: CHUAMRVJSRBRHT-UHFFFAOYSA-N
SMILES: O=CC1=CC(OCC2=CC=CC=C2)=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results