accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOFFF

CAS RN 14618-45-2

IUPAC Name: 2,2,2-trifluoro-1-(1H-indol-3-yl)ethan-1-one
Synonyms: INDOLE3trifluoroacetyl
Molecular Weight (g/mol): 213.16
Molecular Formula: C10H6F3NO
InChi Key: LCMDCXWSHDFQKP-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)C1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)