CAS RN 1462-52-8
IUPAC Name:
3-(dodecylsulfanyl)propanoic acid
Synonyms:
3-(dodecylsulfanyl)propanoic acid
Molecular Weight (g/mol):
274.46
Molecular Formula:
C15H30O2S
InChi Key:
VKLOPQHLJNFYKK-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCSCCC(O)=O