accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHNHOCH3

CAS RN 14620-72-5

IUPAC Name: 1,6-dimethyl hexanediimidate
Synonyms: 1,6-dimethyl hexanediimidate
Molecular Weight (g/mol): 172.23
Molecular Formula: C8H16N2O2
InChi Key: ZLFRJHOBQVVTOJ-UHFFFAOYSA-N
SMILES: COC(=N)CCCCC(=N)OC
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)