accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 146306-75-4

OOHOOHONH(R)CH3

CAS RN 146306-75-4

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-hydroxybutanoic acid
Molecular Weight (g/mol): 341.36
Molecular Formula: C19H19NO5
InChi Key: OYULCCKKLJPNPU-DFDFJRDNSA-N
SMILES: CC(O)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)