accessibility menu, dialog, popup

UserName

Viewing profile as user:
POOHHO(E/Z)

CAS RN 146404-58-2

IUPAC Name: (3-phenylprop-2-en-1-yl)phosphonic acid
Synonyms: 3-phenylprop-2-en-1-ylphosphonic acid
Molecular Weight (g/mol): 198.16
Molecular Formula: C9H11O3P
InChi Key: AIWAKBITCLOITM-UHFFFAOYSA-N
SMILES: OP(O)(=O)CC=CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results