accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14644-55-4

IUPAC Name: caesium(1+) vanadium trioxidandiide
Synonyms: caesium(1+) vanadium trioxidandiide
Molecular Weight (g/mol): 231.85
Molecular Formula: CsO3V
InChi Key: WWENEIGBGOIMMI-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[V].[Cs+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results