accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3ON

CAS RN 146440-15-5

IUPAC Name: 4-[3-(dimethylamino)propoxy]benzonitrile
Synonyms: 4-[3-(dimethylamino)propoxy]benzonitrile
Molecular Weight (g/mol): 204.27
Molecular Formula: C12H16N2O
InChi Key: XYCJVOUBTBJYTL-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results