CAS RN 146447-18-9
IUPAC Name:
1-bromo-4-chloro-2-fluoro-5-methoxybenzene
Synonyms:
1-bromo-4-chloro-2-fluoro-5-methoxybenzene
Molecular Weight (g/mol):
239.47
Molecular Formula:
C7H5BrClFO
InChi Key:
KHRHCBZTZQFNDK-UHFFFAOYSA-N
SMILES:
COC1=CC(Br)=C(F)C=C1Cl
1
–
1
of
1
results
1
5-Bromo-2-chloro-4-fluoroanisole 98.0+%, TCI America™
CAS: 146447-18-9 Molecular Formula: C7H5BrClFO Molecular Weight (g/mol): 239.468 MDL Number: MFCD07784265 InChI Key: KHRHCBZTZQFNDK-UHFFFAOYSA-N Synonym: 1-Bromo-4-chloro-2-fluoro-5-methoxybenzene PubChem CID: 10220378 IUPAC Name: 1-bromo-4-chloro-2-fluoro-5-methoxybenzene SMILES: COC1=CC(=C(C=C1Cl)F)Br