accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14647-23-5

IUPAC Name: [2-(diphenylphosphanyl)ethyl]diphenylphosphane; dichloronickel
Synonyms: diphos; nickel chloride
Molecular Weight (g/mol): 528.02
Molecular Formula: C26H24Cl2NiP2
InChi Key: XXECWTBMGGXMKP-UHFFFAOYSA-L
SMILES: Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (1)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)