accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)PNH2

CAS RN 146476-37-1

IUPAC Name: [(2S)-2-amino-3-methylbutyl]diphenylphosphane
Synonyms: [(2S)-2-amino-3-methylbutyl]diphenylphosphane
Molecular Weight (g/mol): 271.34
Molecular Formula: C17H22NP
InChi Key: ZZLCXURCZWQECA-AGDOHHJYNA-N
SMILES: CC(C)[C@H](N)CP(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results