accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNOH3C

CAS RN 146535-11-7

IUPAC Name: 6,7-dimethoxy-2-phenylquinoxaline
Synonyms: 6,7-dimethoxy-2-phenylquinoxaline
Molecular Weight (g/mol): 266.3
Molecular Formula: C16H14N2O2
InChi Key: QNOXYUNHIGOWNY-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2N=C(C=NC2=C1)C1=CC=CC=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results