accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOFFF

CAS RN 146539-83-5

IUPAC Name: 2-methoxy-5-(trifluoromethyl)benzaldehyde
Synonyms: 2-methoxy-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: WFRURKYDETYZGF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1C=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results