CAS RN 146548-59-6
CAS RN 146548-59-6
IUPAC Name:
1-(2,4,6-trimethoxyphenyl)methanamine hydrochloride
Synonyms:
1-(2,4,6-trimethoxyphenyl)methanamine hydrochloride
Molecular Weight (g/mol):
233.69
Molecular Formula:
C10H16ClNO3
InChi Key:
BLFRMOOGAICNSZ-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC(OC)=C(CN)C(OC)=C1
1
–
1
of
1
results
1
2,4,6-Trimethoxybenzylamine hydrochloride, 98%
CAS: 146548-59-6 Molecular Formula: C10H15NO3·ClH Molecular Weight (g/mol): 233.7 InChI Key: BLFRMOOGAICNSZ-UHFFFAOYSA-N Synonym: 2,4,6-trimethoxybenzylamine hydrochloride,2,4,6-trimethoxyphenyl methanamine hydrochloride,1-2,4,6-trimethoxyphenyl methanamine hydrochloride,pubchem7389,2, 4, 6-trimethoxybenzylamine hydrochloride,acmc-1c5nr,benzenemethanamine, 2,4,6-trimethoxy-, hydrochloride,1-2,4,6-trimethoxyphenyl methanamine-hydrogen chloride 1/1 PubChem CID: 16211914 IUPAC Name: (2,4,6-trimethoxyphenyl)methanamine;hydrochloride SMILES: COC1=CC(=C(C(=C1)OC)CN)OC.Cl