accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14670-94-1

CH3(S)(R)CH3OO

CAS RN 14670-94-1

IUPAC Name: (3R,5S)-3,5-dimethyladamantane-1-carboxylate
Synonyms: (3R,5S)-3,5-dimethyladamantane-1-carboxylate
Molecular Weight (g/mol): 207.29
Molecular Formula: C13H19O2
InChi Key: BSWOQWGHXZTDOO-QGVZIFMBSA-M
SMILES: C[C@@]12CC3C[C@@](C)(C1)CC(C3)(C2)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results