accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 146724-94-9

IUPAC Name: N-[bis(2-azaniumylethyl)amino]-N-nitrosohydroxylamine
Synonyms: N-[bis(2-ammonioethyl)amino]-N-nitrosohydroxylamine
Molecular Weight (g/mol): 165.2
Molecular Formula: C4H15N5O2
InChi Key: HMRRJTFDJAVRMR-UHFFFAOYSA-P
SMILES: [NH3+]CCN(CC[NH3+])N(O)N=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results