accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COS

CAS RN 1468-83-3

IUPAC Name: 1-(thiophen-3-yl)ethan-1-one
Synonyms: 3-acetylthiophene
Molecular Weight (g/mol): 126.17
Molecular Formula: C6H6OS
InChi Key: RNIDWJDZNNVFDY-UHFFFAOYSA-N
SMILES: CC(=O)C1=CSC=C1
Boiling Point 
  • (3)
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (7)
  • (1)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (7)
  • (1)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)