CAS RN 14686-13-6
IUPAC Name:
(2E)-hept-2-ene
Synonyms:
2-heptene (trans)
Molecular Weight (g/mol):
98.19
Molecular Formula:
C7H14
InChi Key:
OTTZHAVKAVGASB-HWKANZROSA-N
SMILES:
CCCC\C=C\C
1
–
1
of
1
results
1
Sigma Aldrich trans-2-Heptene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.