accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 146877-98-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)