accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1469-73-4

IUPAC Name: 4-methyl-1,3,2λ⁴-dioxathiolan-2-one
Synonyms: 4-methyl-1,3,2λ⁴-dioxathiolan-2-one
Molecular Weight (g/mol): 122.14
Molecular Formula: C3H6O3S
InChi Key: SJHAYVFVKRXMKG-UHFFFAOYNA-N
SMILES: CC1COS(=O)O1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)