accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1469-74-5

IUPAC Name: (3-nitrophenyl)(4-nitrophenyl)methanone
Synonyms: (3-nitrophenyl)(4-nitrophenyl)methanone
Molecular Weight (g/mol): 272.22
Molecular Formula: C13H8N2O5
InChi Key: ZEGCOKXUTZGBGN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(=O)C1=CC(=CC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results