accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14690-00-7

IUPAC Name: 2-(benzyloxy)propane-1,3-diol
Synonyms: 2-(benzyloxy)propane-1,3-diol
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: UDIPIOHLDFSMLR-UHFFFAOYSA-N
SMILES: OCC(CO)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results