accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNH3H3CH3CNHO

CAS RN 14691-88-4

IUPAC Name: 1-hydroxy-2,2,6,6-tetramethylpiperidin-4-aminium
Synonyms: 1-hydroxy-2,2,6,6-tetramethylpiperidin-4-aminium
Molecular Weight (g/mol): 173.28
Molecular Formula: C9H21N2O
InChi Key: AYSOUYKQIUYGFR-UHFFFAOYSA-O
SMILES: CC1(C)CC([NH3+])CC(C)(C)N1O
Quantity 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)