accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14694-95-2

IUPAC Name: λ¹-rhodium(1+) tris(triphenylphosphane) chloride
Synonyms: λ¹-rhodium(1+) tris(triphenylphosphine) chloride
Molecular Weight (g/mol): 925.23
Molecular Formula: C54H45ClP3Rh
InChi Key: IXAYKDDZKIZSPV-UHFFFAOYSA-M
SMILES: [Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (7)
Quantity 
  • (5)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (6)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (5)
  • (5)
  • (7)
  • (5)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (6)
  • (3)