accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 146949-07-7

IUPAC Name: 4-propylbenzene-1-sulfonyl chloride
Synonyms: 4-propylbenzenesulfonyl chloride
Molecular Weight (g/mol): 218.7
Molecular Formula: C9H11ClO2S
InChi Key: LEFGAGRZHLNPLS-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)S(Cl)(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results