accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14698-29-4

IUPAC Name: 5-ethyl-8-oxo-2H,5H,8H-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid
Synonyms: ossian uro-alvar acid, oxolinic oxoboi oksaren gramurin uritrate acido ossolico ultibid urotrate
Molecular Weight (g/mol): 261.23
Molecular Formula: C13H11NO5
InChi Key: KYGZCKSPAKDVKC-UHFFFAOYSA-N
SMILES: CCN1C=C(C(O)=O)C(=O)C2=CC3=C(OCO3)C=C12
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results