accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1469816-70-3

IUPAC Name: 3,4-diamino-N'-hydroxybenzene-1-carboximidamide
Synonyms: 3,4-diamino-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 166.18
Molecular Formula: C7H10N4O
InChi Key: OPOKBHCWRPVYFH-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC(N)=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results