accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 146982-30-1

OOHOOONH(S)CH2

CAS RN 146982-30-1

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[4-(prop-2-en-1-yloxy)phenyl]propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[4-(prop-2-en-1-yloxy)phenyl]propanoic acid
Molecular Weight (g/mol): 443.5
Molecular Formula: C27H25NO5
InChi Key: AQUXDQFCBLRIRE-VWLOTQADSA-N
SMILES: OC(=O)[C@H](CC1=CC=C(OCC=C)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)