accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CNNONH(S)OHONHOO(E/Z)

CAS RN 146998-27-8

IUPAC Name: (2S)-6-[(4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}phenyl)formamido]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-6-[(4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}phenyl)formamido]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 619.72
Molecular Formula: C36H37N5O5
InChi Key: FPOPWTDBGMLRNG-YGFIFORBNA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)