accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHOI

CAS RN 147-58-0

IUPAC Name: 2-[(2-iodophenyl)formamido]acetic acid
Synonyms: ortho-iodohippurate
Molecular Weight (g/mol): 305.07
Molecular Formula: C9H8INO3
InChi Key: CORFWQGVBFFZHF-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)C1=CC=CC=C1I
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results