CAS RN 147-71-7
IUPAC Name:
2,3-dihydroxybutanedioic acid
Synonyms:
(.+-.)-tartaric acid|D-(−)-tartaric acid
Molecular Weight (g/mol):
150.09
Molecular Formula:
C4H6O6
InChi Key:
FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES:
OC(C(O)C(O)=O)C(O)=O
1
–
6
of
6
results
2
Promotions Available
D(-)-Tartaric acid, 99%
D(-)-Tartaric acid, 99%
3
Promotions Available
D-(-)-Tartaric acid, 99%
D-(-)-Tartaric acid, 99%
4