accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1470-94-6

IUPAC Name: 2,3-dihydro-1H-inden-5-ol
Synonyms: 5-indanol
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: PEHSSTUGJUBZBI-UHFFFAOYSA-N
SMILES: OC1=CC=C2CCCC2=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)