accessibility menu, dialog, popup

UserName

Viewing profile as user:
HN

CAS RN 1470-99-1

IUPAC Name: octahydro-1H-isoindole
Synonyms: octahydro-1H-isoindole
Molecular Weight (g/mol): 125.22
Molecular Formula: C8H15N
InChi Key: ODSNARDHJFFSRH-UHFFFAOYNA-N
SMILES: C1NCC2CCCCC12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results